4-Fluorobenzo[b]thiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

044604-500MG 044604-1G 044604-5G
101866-670EA 175.38 USD
101866-670 101866-672 101866-674
4-Fluorobenzo[b]thiophene-2-carboxylic acid ≥95%
4-Fluorobenzo[b]thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅FO₂S
MW: 196.1982 g/mol
Melting Pt: 217-220°C
MDL Number: MFCD03425782
CAS Number: 310466-37-6

Order Now

Specification Test Results

Melting point 217-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR