Methyl-4-fluorobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-fluorobenzo[b]thiophene-2-carboxylate

044603-500MG 044603-1G 044603-5G
101866-664EA 278.46 USD
101866-664 101866-666 101866-668
Methyl-4-fluorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 4-fluorobenzo[b]thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₇FO₂S
MW: 210.2248 g/mol
Melting Pt: 96-98°C
MDL Number: MFCD06200964
CAS Number: 220180-55-2

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR