3-Imidazo[2,1-b][1,3]benzothiazol-2-ylaniline ≥95%

Supplier: Matrix Scientific

044599-500MG 044599-1G
101866-652EA 556.92 USD
101866-652 101866-654
3-Imidazo[2,1-b][1,3]benzothiazol-2-ylaniline ≥95%
3-Imidazo[2,1-b][1,3]benzothiazol-2-ylaniline

Hazard: Irritant

Formula: C₁₅H₁₁N₃S
Melting Pt: 169…171 °C
MDL Number: MFCD03791217
CAS Number: 79890-07-6

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR