5-Methyl-2-furaldehyde oxime ≥95%
Supplier: Matrix Scientific
Synonyms:
(E)-N-[(5-Methylfuran-2-yl)methylidene]hydroxylamine
044597-500MG
044597-1G
044597-5G
101866-646EA
230.77
USD
101866-646
101866-648
101866-650
5-Methyl-2-furaldehyde oxime ≥95%
5-Methyl-2-furaldehyde oxime
Hazard: Irritant
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Melting Pt: 102…104 °C |
MDL Number:
MFCD00482440 CAS Number: 32750-36-0 |
Specification Test Results
Melting point | 102-104°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...