3-(2-Quinoxalinyl)aniline ≥95%

Supplier: Matrix Scientific

044595-500MG 044595-1G 044595-5G
101866-636EA 375.38 USD
101866-636 101866-638 101866-640
3-(2-Quinoxalinyl)aniline ≥95%
3-(2-Quinoxalinyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₁N₃
Melting Pt: 164…166 °C
MDL Number: MFCD02737921
CAS Number: 432004-76-7

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR