7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

044594-500MG 044594-1G
101866-632EA 278.46 USD
101866-632 101866-634
7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine ≥95%
7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
Melting Pt: 178…180 °C
MDL Number: MFCD00438465
CAS Number: 54970-96-6

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR