2,4-Dichloro-5-[(3-fluorobenzyl)oxy]aniline ≥95%

Supplier: Matrix Scientific

044589-500MG 044589-1G 044589-5G
101866-604EA 556.92 USD
101866-604 101866-606 101866-608
2,4-Dichloro-5-[(3-fluorobenzyl)oxy]aniline ≥95%
2,4-Dichloro-5-[(3-fluorobenzyl)oxy]aniline

Hazard: Irritant

Formula: C₁₃H₁₀Cl₂FNO
Melting Pt: 84-86°C
MDL Number: MFCD03425777
CAS Number: 439095-33-7

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR