2-(Acetylamino)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

044587-500MG 044587-1G 044587-5G
101866-592EA 181.54 USD
101866-592 101866-594 101866-596
2-(Acetylamino)benzenecarboxamide ≥95%
2-(Acetylamino)benzenecarboxamide

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
Melting Pt: 182…184 °C
MDL Number: MFCD00157809
CAS Number: 33809-77-7

Order Now

Specification Test Results

Melting point 182-184°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR