Ethyl-5-amino-1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044584-500MG 044584-1G 044584-5G
101866-574EA 181.54 USD
101866-574 101866-576 101866-578
Ethyl-5-amino-1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-5-amino-1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁Cl₂N₃O₂
Melting Pt: 175…178 °C
MDL Number: MFCD01569432
CAS Number: 83279-66-7

Order Now

Specification Test Results

Melting point 175-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR