N-(2,4-Dichlorophenyl)-2-(4-hydroxyphenyl)acetamide ≥95%

Supplier: Matrix Scientific

044583-500MG 044583-1G
101866-570EA 181.54 USD
101866-570 101866-572
N-(2,4-Dichlorophenyl)-2-(4-hydroxyphenyl)acetamide ≥95%
N-(2,4-Dichlorophenyl)-2-(4-hydroxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₄H₁₁Cl₂NO₂
Melting Pt: 136…140 °C
MDL Number: MFCD03425774
CAS Number: 439095-30-4

Order Now

Specification Test Results

Melting point 136-140°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR