3-Hydroxy-2-phenyl-4(1H)-quinolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-phenylquinolin-4(1H)-one

044582-500MG 044582-1G
101866-566EA 278.46 USD
101866-566 101866-568
3-Hydroxy-2-phenyl-4(1H)-quinolinone ≥95%
3-Hydroxy-2-phenyl-4(1H)-quinolinone

Hazard: Irritant

Formula: C₁₅H₁₁NO₂
MW: 237.26 g/mol
Melting Pt: 268…270 °C
MDL Number: MFCD00451324
CAS Number: 31588-18-8

Order Now

Specification Test Results

Melting point 268-270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR