1-(1,3-Benzodioxol-5-yl)-3-(dimethylamino)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

044581-500MG 044581-1G 044581-5G
101866-560EA 181.54 USD
101866-560 101866-562 101866-564
1-(1,3-Benzodioxol-5-yl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(1,3-Benzodioxol-5-yl)-3-(dimethylamino)-2-propen-1-one

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
Melting Pt: 124…126 °C
MDL Number: MFCD06659552
CAS Number: 154257-92-8

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR