Methyl-4-(3-amino-4-nitrophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044580-500MG 044580-1G
101866-556EA 346.15 USD
101866-556 101866-558
Methyl-4-(3-amino-4-nitrophenoxy)benzenecarboxylate ≥95%
Methyl-4-(3-amino-4-nitrophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₅
MW: 288.26 g/mol
Melting Pt: 175 °C
MDL Number: MFCD03425772
CAS Number: 439095-28-0

Order Now

Specification Test Results

Melting point 175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR