4-(2-Pyridinyl)-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-4-(pyridin-2-yl)-1H-pyrazole

044578-500MG 044578-1G 044578-5G
101866-546EA 278.46 USD
101866-546 101866-548 101866-550
4-(2-Pyridinyl)-1H-pyrazol-5-amine ≥95%
4-(2-Pyridinyl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₈H₈N₄
MW: 160.18 g/mol
Melting Pt: 120…121 °C
MDL Number: MFCD03617434
CAS Number: 493038-87-2

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR