2-[5-Nitro-2-oxo-1,3-benzoxazol-3(2H)-yl]acetamide ≥95%

Supplier: Matrix Scientific

044577-500MG 044577-1G
101866-542EA 375.38 USD
101866-542 101866-544
2-[5-Nitro-2-oxo-1,3-benzoxazol-3(2H)-yl]acetamide ≥95%
2-[5-Nitro-2-oxo-1,3-benzoxazol-3(2H)-yl]acetamide

Hazard: Irritant

Formula: C₉H₇N₃O₅
Melting Pt: 234…235 °C
MDL Number: MFCD03425771
CAS Number: 439095-26-8

Order Now

Specification Test Results

Melting point 234-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR