2-Oxo-2-phenylethyl-2-amino-3,5-dichlorobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

044576-500MG 044576-1G 044576-5G
101866-536EA 181.54 USD
101866-536 101866-538 101866-540
2-Oxo-2-phenylethyl-2-amino-3,5-dichlorobenzenecarboxylate ≥95%
2-Oxo-2-phenylethyl-2-amino-3,5-dichlorobenzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₁Cl₂NO₃
Melting Pt: 180…182 °C
MDL Number: MFCD06557573
CAS Number: 748158-74-9

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR