1-Bromo-4-(tert-butoxy)benzene ≥95%

Supplier: Matrix Scientific

044575-500MG 044575-1G 044575-5G
101866-530EA 375.38 USD
101866-530 101866-532 101866-534
1-Bromo-4-(tert-butoxy)benzene ≥95%
1-Bromo-4-(tert-butoxy)benzene

Hazard: Irritant

Formula: C₁₀H₁₃BrO
Melting Pt: 32…33 °C
MDL Number: MFCD00792676
CAS Number: 60876-70-2

Order Now

Specification Test Results

Melting point 32-33°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR