3-(tert-Butoxy)-6-chloropyridazine ≥95%

Supplier: Matrix Scientific

044573-500MG 044573-1G
101866-520EA 230.77 USD
101866-520 101866-522
3-(tert-Butoxy)-6-chloropyridazine ≥95%
3-(tert-Butoxy)-6-chloropyridazine

Hazard: Irritant

Formula: C₈H₁₁ClN₂O
MW: 186.6400 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD03425769
CAS Number: 17321-24-3

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR