6-Hydroxy-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Cyano-4-methyl-2,6-pyridinediol, 3-Cyano-2,6-dihydroxy-4-picoline, 2,6-Dihydroxy-4-methylnicotinonitrile, 3-Cyano-6-hydroxy-4-methyl-2-pyridone

044572-500MG 044572-1G 044572-5G
101866-514EA 136.92 USD
101866-514 101866-516 101866-518
6-Hydroxy-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥95%
6-Hydroxy-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Hazard: Irritant

Formula: C₇H₆N₂O₂
MW: 150.1347 g/mol
Melting Pt: 315…320 °C
MDL Number: MFCD00043831
CAS Number: 5444-02-0

Order Now

Specification Test Results

Melting point 315-320°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR