1-((4-Nitrophenyl)sulfonyl)-4-pyridinecarboxylic acid ≥95%

Supplier: Matrix Scientific

044565-500MG 044565-1G 044565-5G
101866-488EA 278.46 USD
101866-488 101866-490 101866-492
1-((4-Nitrophenyl)sulfonyl)-4-pyridinecarboxylic acid ≥95%
1-((4-Nitrophenyl)sulphonyl)-4-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₆S
Melting Pt: 248-250°C
MDL Number: MFCD01045658
CAS Number: 331828-49-0

Order Now

Specification Test Results

Melting point 248-250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR