2-(Phenylsulfonyl)-3-pyridinamine ≥95%

Supplier: Matrix Scientific

044557-500MG 044557-1G 044557-5G
101866-466EA 375.38 USD
101866-466 101866-468 101866-470
2-(Phenylsulfonyl)-3-pyridinamine ≥95%
2-(Phenylsulfonyl)-3-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂S
Melting Pt: 172…174 °C
MDL Number: MFCD02186480
CAS Number: 444087-50-7

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR