3-Methoxy-3-oxo-1-propenyl4-aminobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

044555-500MG 044555-1G 044555-5G
101866-454EA 181.54 USD
101866-454 101866-456 101866-458
3-Methoxy-3-oxo-1-propenyl4-aminobenzenecarboxylate ≥95%
3-Methoxy-3-oxo-1-propenyl4-aminobenzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
Melting Pt: 186…188 °C
MDL Number: MFCD03001267
CAS Number: 400878-23-1

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR