3-Methoxy-3-oxo-1-propenyl-4-(trifluoromethyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044553-500MG 044553-1G 044553-5G
101866-442EA 375.38 USD
101866-442 101866-444 101866-446
3-Methoxy-3-oxo-1-propenyl-4-(trifluoromethyl)benzenecarboxylate ≥95%
3-Methoxy-3-oxo-1-propenyl-4-(trifluoromethyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₉F₃O₄
MW: 274.2 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD03001265
CAS Number: 400878-21-9

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR