1-(4-Cyanophenyl)piperazine ≥95%

Supplier: Matrix Scientific

044552-1G 044552-5G 044552-25G
101866-436EA 29.23 USD
101866-436 101866-438 101866-440
1-(4-Cyanophenyl)piperazine ≥95%
1-(4-Cyanophenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₃N₃
Melting Pt: 86…88 °C
MDL Number: MFCD01320890
CAS Number: 68104-63-2

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR