3-Methoxy-3-oxo-1-propenyl4-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

044551-500MG 044551-1G 044551-5G
101866-430EA 300 USD
101866-430 101866-432 101866-434
3-Methoxy-3-oxo-1-propenyl4-nitrobenzenecarboxylate ≥95%
3-Methoxy-3-oxo-1-propenyl4-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₆
Melting Pt: 152…154 °C
MDL Number: MFCD03001264
CAS Number: 1104461-74-6

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR