3,4-Dibromo-5-phenyl-2(5H)-furanone ≥95%

Supplier: Matrix Scientific

044547-500MG 044547-1G 044547-5G
101866-414EA 278.46 USD
101866-414 101866-416 101866-418
3,4-Dibromo-5-phenyl-2(5H)-furanone ≥95%
3,4-Dibromo-5-phenyl-2(5H)-furanone

Hazard: Irritant

Formula: C₁₀H₆Br₂O₂
Melting Pt: 74…76 °C
MDL Number: MFCD03001263
CAS Number: 72857-86-4

Order Now

Specification Test Results

Melting point 74-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR