2-Amino-1-(4-methoxybenzyl)-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

044544-500MG 044544-1G 044544-5G
101866-398EA 278.46 USD
101866-398 101866-400 101866-402
2-Amino-1-(4-methoxybenzyl)-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-1-(4-methoxybenzyl)-4,5-dimethyl-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₁₅H₁₇N₃O
Melting Pt: 142…144 °C
MDL Number: MFCD03001262
CAS Number: 451485-75-9

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR