2-(2-Formylphenoxy)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

044539-500MG 044539-1G
101866-380EA 556.92 USD
101866-380 101866-382
2-(2-Formylphenoxy)benzenecarbonitrile ≥95%
2-(2-Formylphenoxy)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₄H₉NO₂
Melting Pt: 72…74 °C
MDL Number: MFCD02082410
CAS Number: 451485-74-8

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR