4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one ≥95%
Supplier: Matrix Scientific
044537-500MG
044537-1G
044537-5G
101866-374EA
1109.23
USD
101866-374
101866-376
101866-378
4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one ≥95%
4-[2-(4-Chloro-3-fluorophenoxy)phenyl]-3-buten-2-one
Hazard: Irritant
Formula:
C₁₆H₁₂ClFO₂ Melting Pt: 68…70 °C |
MDL Number:
MFCD02091020 CAS Number: 451485-73-7 |
Specification Test Results
Melting point | 68-70°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...