4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one ≥95%

Supplier: Matrix Scientific

044536-500MG 044536-1G 044536-5G
101866-368EA 921.54 USD
101866-368 101866-370 101866-372
4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one ≥95%
4-[2-(4-Bromo-2-fluorophenoxy)phenyl]-3-buten-2-one

Hazard: Irritant

Formula: C₁₆H₁₂BrFO₂
Melting Pt: 83…85 °C
MDL Number: MFCD02091019
CAS Number: 451485-72-6

Order Now

Specification Test Results

Melting point 83-85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR