2-(4,5-Dihydro-1H-imidazol-2-yl)thiophenol ≥95%

Supplier: Matrix Scientific

044535-500MG 044535-1G 044535-5G
101866-362EA 104.62 USD
101866-362 101866-364 101866-366
2-(4,5-Dihydro-1H-imidazol-2-yl)thiophenol ≥95%
2-(4,5-Dihydro-1H-imidazol-2-yl)thiophenol

Hazard: Irritant

Formula: C₉H₁₀N₂S
Melting Pt: 210°C
MDL Number: MFCD01815126
CAS Number: 53440-31-6

Order Now

Specification Test Results

Melting point 210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR