3-Quinuclidinone oxime ≥95%

Supplier: Matrix Scientific

044534-500MG 044534-1G
101866-358EA 375.38 USD
101866-358 101866-360
3-Quinuclidinone oxime ≥95%
3-Quinuclidinone oxime

Hazard: Irritant

Formula: C₇H₁₂N₂O
Melting Pt: 223…225 °C
MDL Number: MFCD03768424
CAS Number: 35423-17-7

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR