N-(3-Chlorophenyl)-N'-(2-hydroxy-1-phenylethyl)urea ≥95%

Supplier: Matrix Scientific

044529-500MG 044529-1G 044529-5G
101866-344EA 747.69 USD
101866-344 101866-346 101866-348
N-(3-Chlorophenyl)-N'-(2-hydroxy-1-phenylethyl)urea ≥95%
N-(3-Chlorophenyl)-N'-(2-hydroxy-1-phenylethyl)urea

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₂
MW: 290.75 g/mol
Melting Pt: 182…184 °C
MDL Number: MFCD01316023
CAS Number: 338750-54-2

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR