Methyl-2-(acetylamino)-5-chloro-3-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044527-500MG 044527-1G 044527-5G
101866-338EA 346.15 USD
101866-338 101866-340 101866-342
Methyl-2-(acetylamino)-5-chloro-3-thiophenecarboxylate ≥95%
Methyl-2-(acetylamino)-5-chloro-3-thiophenecarboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₃S
Melting Pt: 109…111 °C
MDL Number: MFCD01316143
CAS Number: 22288-82-0

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR