Methyl-2-(acetylamino)-3-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044526-500MG 044526-1G 044526-5G
101866-332EA 269.23 USD
101866-332 101866-334 101866-336
Methyl-2-(acetylamino)-3-thiophenecarboxylate ≥95%
Methyl-2-(acetylamino)-3-thiophenecarboxylate

Hazard: Irritant

Formula: C₈H₉NO₃S
Melting Pt: 103…105 °C
MDL Number: MFCD00728968
CAS Number: 22288-81-9

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR