5-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]-1,3,4-thiadiazol-2-amine ≥95%

Supplier: Matrix Scientific

044524-500MG 044524-1G 044524-5G
101866-326EA 181.54 USD
101866-326 101866-328 101866-330
5-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]-1,3,4-thiadiazol-2-amine ≥95%
5-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₆H₆F₃N₃S₂
Melting Pt: 160…162 °C
MDL Number: MFCD01443829
CAS Number: 145070-01-5

Order Now

Specification Test Results

Melting point 160-162°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR