Methyl-4-(2-chlorophenyl)-2,4-dioxobutanoate ≥95%

Supplier: Matrix Scientific

044522-500MG 044522-1G 044522-5G
101866-314EA 81.54 USD
101866-314 101866-316 101866-318
Methyl-4-(2-chlorophenyl)-2,4-dioxobutanoate ≥95%
Methyl-4-(2-chlorophenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₁H₉ClO₄
Melting Pt: 78…80 °C
MDL Number: MFCD03001316
CAS Number: 451485-68-0

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR