4-Chloro-N-(5-iodo-2-pyridinyl)butanamide ≥95%

Supplier: Matrix Scientific

044517-500MG 044517-1G 044517-5G
101866-290EA 375.38 USD
101866-290 101866-292 101866-294
4-Chloro-N-(5-iodo-2-pyridinyl)butanamide ≥95%
4-Chloro-N-(5-iodo-2-pyridinyl)butanamide

Hazard: Irritant

Formula: C₉H₁₀ClIN₂O
Melting Pt: 132…134 °C
MDL Number: MFCD01315195
CAS Number: 338748-92-8

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR