1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Chloro-6-fluorobenzyl)-5-cyano-6-methyl-1,2,3,4-tetrahydropyridin-2-one

044514-500MG 044514-1G
101866-276EA 375.38 USD
101866-276 101866-278
1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₂ClFN₂O
MW: 278.72 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD00975042
CAS Number: 338748-74-6

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR