1-(Mesitylmethyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

044513-500MG 044513-1G 044513-5G
101866-270EA 375.38 USD
101866-270 101866-272 101866-274
1-(Mesitylmethyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
1-(Mesitylmethyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₇H₂₀N₂O
Melting Pt: 150…152 °C
MDL Number: MFCD00975041
CAS Number: 338748-73-5

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR