1-(2,4-Dichlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

044512-500MG 044512-1G 044512-5G
101866-264EA 375.38 USD
101866-264 101866-266 101866-268
1-(2,4-Dichlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
1-(2,4-Dichlorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₂Cl₂N₂O
Melting Pt: 144…146 °C
MDL Number: MFCD00975040
CAS Number: 338748-72-4

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR