1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

044511-500MG 044511-1G
101866-260EA 375.38 USD
101866-260 101866-262
1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
1-(4-Fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₃FN₂O
MW: 244.27 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00975039
CAS Number: 338748-69-9

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR