2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

044510-500MG 044510-1G 044510-5G
101866-254EA 181.54 USD
101866-254 101866-256 101866-258
2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol ≥95%
2-{4-[2-Nitro-4-(trifluoromethyl)phenyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₁₆F₃N₃O₃
Melting Pt: 108…110 °C
MDL Number: MFCD01173874
CAS Number: 330633-81-3

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR