2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol ≥95%

Supplier: Matrix Scientific

044509-500MG 044509-1G 044509-5G
101866-248EA 60 USD
101866-248 101866-250 101866-252
2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol ≥95%
2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
Melting Pt: 97…99 °C
MDL Number: MFCD00511837
CAS Number: 5521-38-0

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR