Ethyl-2-methyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydro-3-quinolinecarboxylate ≥95%

Supplier: Matrix Scientific

044508-500MG 044508-1G 044508-5G
101866-242EA 278.46 USD
101866-242 101866-244 101866-246
Ethyl-2-methyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydro-3-quinolinecarboxylate ≥95%
Ethyl-2-methyl-5-oxo-4-phenyl-1,4,5,6,7,8-hexahydro-3-quinolinecarboxylate

Hazard: Irritant

Formula: C₁₉H₂₁NO₃
Melting Pt: 260…262 °C
MDL Number: MFCD00619417
CAS Number: 33323-03-4

Order Now

Specification Test Results

Melting point 260-262°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR