Methyl-2-(methylsulfonyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(methylsulphonyl)benzoate

044504-500MG 044504-1G 044504-5G
101866-218EA 200 USD
101866-218 101866-220 101866-222
Methyl-2-(methylsulfonyl)benzenecarboxylate ≥95%
Methyl-2-(methylsulfonyl)benzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD03001211
CAS Number: 199657-02-8

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR