4-Chlorobenzyl(6-chloro-2-phenyl-4-pyrimidinyl)methyl sulfide ≥95%
Supplier: Matrix Scientific
044502-500MG
044502-1G
044502-5G
101866-206EA
375.38
USD
101866-206
101866-208
101866-210
4-Chlorobenzyl(6-chloro-2-phenyl-4-pyrimidinyl)methyl sulfide ≥95%
4-Chlorobenzyl(6-chloro-2-phenyl-4-pyrimidinyl)methyl sulfide
Hazard: Irritant
Formula:
C₁₈H₁₄Cl₂N₂S MW: 361.3 g/mol Melting Pt: 116…117 °C |
MDL Number:
MFCD00974415 CAS Number: 339279-19-5 |
Specification Test Results
Melting point | 116-117°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...