6-(Methoxymethyl)-2-phenyl-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
044500-500MG
044500-1G
044500-5G
101866-194EA
278.46
USD
101866-194
101866-196
101866-198
6-(Methoxymethyl)-2-phenyl-4-pyrimidinol ≥95%
6-(Methoxymethyl)-2-phenyl-4-pyrimidinol
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₂ Melting Pt: 183…185 °C |
MDL Number:
MFCD00269583 CAS Number: 339278-89-6 |
Specification Test Results
Melting point | 183-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...