Methyl-5-phenyl-1,2,4-oxadiazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

044491-500MG 044491-1G 044491-5G
101866-148EA 347.69 USD
101866-148 101866-150 101866-152
Methyl-5-phenyl-1,2,4-oxadiazole-3-carboxylate ≥95%
Methyl-5-phenyl-1,2,4-oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
Melting Pt: 113…116 °C
MDL Number: MFCD00974530
CAS Number: 37384-61-5

Order Now

Specification Test Results

Melting point 113-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR