2',4'-Dichloro-5'-hydroxyacetanilide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2,4-Dichloro-5-hydroxyphenyl)acetamide

044489-500MG 044489-1G 044489-5G
101866-136EA 278.46 USD
101866-136 101866-138 101866-140
2',4'-Dichloro-5'-hydroxyacetanilide ≥95%
2',4'-Dichloro-5'-hydroxyacetanilide

Hazard: Irritant

Formula: C₈H₇Cl₂NO₂
MW: 220.0527 g/mol
Melting Pt: 234-236°C
MDL Number: MFCD00186236
CAS Number: 67669-19-6

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR